missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric acid, pure, 25% solution in water

CAS: 7647-01-0 | ClH | 36.46 g/mol

$68.13 - $127.59

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
SMILES Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC387790010
SDS View Documents
Thermo Scientific Chemicals
387790010
1 L
Each for $68.13
Only null left
 
AC387790025
SDS View Documents
Thermo Scientific Chemicals
387790025
2.5 L
Each for $127.59
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7647-01-0
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
Cl
ClH
MFCD00011324 MFCD00792839
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
CHEBI:17883

Specifications

Colorless
7647-01-0 , 7732-18-5
1.1200g/mL
Glass bottle
1.12
ClH
HCl
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
VEXZGXHMUGYJMC-UHFFFAOYSA-N
chlorane
313
36.45
Pure
-70.0°C
107.0°C
1ppm max.
0.002% max.
1 L
Liquid
MFCD00011324 MFCD00792839
Solubility in water: soluble.
Cl
36.46
CHEBI:17883
≥25.0%
Hydrochloric acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation. Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

Danger

SDS
Documents

Documents

RUO – Research Use Only