missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid (Optima™), Fisher Chemical™

Catalog No. p-20174
Change view
Click to view available options
Quantity:
250 mL
500 mL
1 L
2 L
Packaging:
FEP Bottle
PFA Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A466250 250 mL PFA Bottle
A466500 500 mL PFA Bottle
A4661 1 L PFA Bottle
A4662 2 L FEP Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. A466250 Supplier Seastar Chemicals Inc Supplier No. S020401TFIF01
Only null left

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

pH 1
Color Colorless
Melting Point -35°C
Boiling Point 57°C
Density 1.18g/cm³
Packaging PFA Bottle
Vapor Pressure 125mbar at 20°C
Certificate of Analysis Certificate of Lot Analysis included with each bottle.
Quantity 250 mL
Physical Form Liquid
UN Number 1789
Formula Weight 36.46
Percent Purity 32 to 35% (HCl, w/w)
Grade OPTIMA
Chemical Name or Material Hydrochloric Acid
Show More Show Less
ShelfLife Three years
DOTInformation DOT Class 8, : Corrosive