missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid (Optima™), Fisher Chemical™
Supplier: Fisher Chemical S020401TFIF07
Specifications
| 1 | |
| Colorless | |
| 7647-01-0 | |
| 1.18g/cm³ | |
| 125mbar at 20°C | |
| 2 L | |
| Liquid | |
| 1789 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| hydrogen chloride | |
| 313 | |
| 36.46 | |
| OPTIMA |
| Hydrochloric Acid | |
| -35°C | |
| 57°C | |
| FEP Bottle | |
| Certificate of Lot Analysis included with each bottle. | |
| ClH | |
| MFCD00011324 MFCD00792839 | |
| hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
| Cl | |
| 36.46 | |
| CHEBI:17883 | |
| 32 to 35% (HCl, w/w) |
Chemical Identifiers
| 7647-01-0 | |
| 36.46 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| 313 | |
| hydrogen chloride |
| ClH | |
| MFCD00011324 MFCD00792839 | |
| hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
| CHEBI:17883 | |
| Cl |
Safety and Handling
ShelfLife : 3 years
DOTInformation : DOT Class 8, : Corrosive