missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid (NF/FCC), Fisher Chemical

Catalog No. A481212
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A481212 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A481212 Supplier Thermo Fisher Scientific Formerly Fairlawn Supplier No. FLA481212
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 7647-01-0 , 7732-18-5
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

pH 1
Color Colorless
Melting Point -66°C
Boiling Point 83°C
Density 1.27 (Air = 1.0)
Heavy Metals (as Pb) 5ppm max. (NF)
Identification Pass Test (FCC), Pass Test (FCC/NF), Pass Test (NF)
Non_volatile Matter 0.5% max. (FCC)
Packaging Glass Bottle
Reducing Substances 0.007% max. (FCC)
Residue after Ignition 0.008% max. (NF)
Specific Gravity 1.1798 to 1.1944 (15.6°C/60°F)
Vapor Pressure 125mbar at 20°C
Quantity 2.5 L
Physical Form Liquid
UN Number 1789
Formula Weight 36.46
Percent Purity 36.5 to 38.0% (NF)
Grade FCC/NF
Chemical Name or Material Hydrochloric Acid
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Causes burns by all exposure routes. Use only under a chemical fume hood. Wear personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Do not induce vomiting. Do not induce vomiting. Call a physician or Poison Control Center immediately.

NFPA

Health:3
Flammability:0
Instability:1
DOTInformation Class 8: Corrosive