missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, Certified, 6.0N ±0.1N (6.0M), LabChem™

Catalog No. p-5379556
Change view
Click to view available options
Quantity:
1 L
2.5 L
10 L
20 L
Catalog No. Quantity
LC153702 1 L
LC153703 2.5 L
LC153706 10 L
LC153705 20 L
4 options

Catalog No. LC153702

Supplier: LabChem LC153702

Only null left

Chemical Identifiers

CAS 7647-01-0 , 7732-18-5
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

Color Colorless
Concentration 6.0N ±0.1N (6.0M)
CAS Max % 79.92, 20.08
Identification Passes Test
Packaging Amber Glass
Traceability to NIST Traceable to NIST
Decomposition Information Hydrogen chloride; Thermal decomposition generates: Corrosive vapors
Quantity 1 L
Physical Form Liquid
Linear Formula HCl
UN Number UN1789
Solubility Information Soluble in water
Formula Weight 36.46
Grade Certified
Chemical Name or Material Hydrochloric Acid
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Do not breathe mist, vapors, spray.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Store in a well-ventilated place.
Keep container tightly closed.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature