missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, Certified, 6.0N ±0.1N (6.0M), LabChem™
Specifications
Specifications
| Color | Colorless |
| Concentration | 6.0N ± 0.1N |
| CAS Max % | 79.92, 20.08 |
| Identification | Passes Test |
| Packaging | HDPE Bottle |
| Traceability to NIST | Traceable to NIST |
| Decomposition Information | Hydrogen chloride; Thermal decomposition generates: Corrosive vapors |
| Quantity | 2.5 L |
| Physical Form | Liquid |
| Linear Formula | HCl |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement Do not breathe mist, vapors, spray. Use only outdoors or in a well-ventilated area. Wear protective gloves, protective clothing, eye protection, face protection. Wash exposed skin thoroughly after handling. Store |
| Recommended Storage | Room Temperature |