missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 10 g/L HCl, Ricca Chemical

Catalog No. R35630001A
Change view
Click to view available options
Quantity:
1 L
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R35630001A 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R35630001A Supplier Ricca Chemical Company Supplier No. R35630001A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Hydrochloric acid solution for general laboratory use.

Chemical Identifiers

CAS 7732-18-5 , 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid
PubChem CID 313
ChEBI CHEBI:23116
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

pH <1
Appearance Clear colorless liquid
Odor Odorless to slightly pungent
Packaging Natural Poly Bottle
Quantity 1 L
Physical Form Liquid
Solubility Information Solubility in Water: Infinite
Grade Laboratory
Chemical Name or Material Hydrochloric Acid
Recommended Storage General