missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, ACS Reagent Grade, Ricca Chemical
Supplier: Ricca Chemical Company RABH00101C
Specifications
| Hydrochloric Acid | |
| 7647-01-0 | |
| 100.0 | |
| Amber Glass Bottle | |
| 1 L | |
| Liquid | |
| hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
| Cl | |
| 36.46 | |
| CHEBI:17883 | |
| ACS |
| Colorless to Green-Yellow | |
| 98.0 | |
| max. 1 ppm | |
| max. 5 ppm | |
| ClH | |
| MFCD00011324 MFCD00792839 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| hydrogen chloride | |
| 313 | |
| 36.5 to 38.0% |
Chemical Identifiers
| 7647-01-0 | |
| 36.46 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| CHEBI:17883 | |
| Cl |
| ClH | |
| MFCD00011324 MFCD00792839 | |
| 313 | |
| hydrogen chloride |
Safety and Handling
ShelfLife : 36 months