missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, ACS Reagent Grade, Ricca Chemical

Catalog No. p-3251792
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Packaging:
Amber Glass Bottle
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RABH0010500 500 mL Glass Bottle
RABH00101C 1 L Amber Glass Bottle
RABH001025 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. RABH0010500 Supplier Ricca Chemical Company Supplier No. RABH0010500D1
Only null left

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

Color Colorless to Green-Yellow
CAS Min % 98.0
CAS Max % 100.0
Heavy Metals (as Pb) max. 1 ppm
Packaging Glass Bottle
Residue after Ignition max. 5 ppm
Quantity 500 mL
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
Percent Purity 36.5 to 38.0%
Grade ACS
Chemical Name or Material Hydrochloric Acid
Show More Show Less
ShelfLife 36 months