missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, ACS Reagent, 37%, Honeywell Fluka™

ACS Reagent, 37%

Supplier:  Honeywell Inc 2581484X4L

 View more versions of this product

Catalog No. 6000709

Only null left
Explore available promotions

Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Specifications

Hydrochloric Acid
37%
≤5ppm
4 x 4 L
MFCD00011324 MFCD00792839
1098214
VEXZGXHMUGYJMC-UHFFFAOYSA-N
chlorane
313
36.46g/mol
7647-01-0
1.2 g/mL at 25°C
3.23psi (21.1°C), 7.93psi (37.7°C)
ClH
1789
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
Cl
36.46
CHEBI:17883
ACS Reagent

Chemical Identifiers

7647-01-0
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
Cl
ClH
MFCD00011324 MFCD00792839
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
CHEBI:17883
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P260-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338

H290-H314-H335

EINECSNumber : 231-595-7

RTECSNumber : MW4025000

SDS
Documents

Documents

Promotions

Promotions