missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, ACS Reagent, 37%, Honeywell Fluka™

Catalog No. p-7071945
Change view
Click to view available options
Quantity:
500 mL
4 L
6 x 500 mL
4 x 4 L
Density:
1.2 g/mL at 25°C
Catalog No. Quantity Density
6000710 500 mL 1.2 g/mL at 25°C
6000712 6 x 500 mL 1.2 g/mL at 25°C
6000708 4 L 1.2 g/mL at 25°C
6000709 4 x 4 L 1.2 g/mL at 25°C
4 options

Catalog No. 6000710

Supplier: Honeywell Inc 258148500ML

Only null left

ACS Reagent, 37%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
SMILES Cl

Specifications

Concentration 37%
Density 1.2 g/mL at 25°C
Residue after Ignition ≤5ppm
Vapor Pressure 3.23psi (21.1°C), 7.93psi (37.7°C)
Quantity 500 mL
UN Number 1789
Beilstein 1098214
IUPAC Name hydrogen chloride
Formula Weight 36.46g/mol
Grade ACS Reagent
Chemical Name or Material Hydrochloric Acid
Show More Show Less
Hazard Statement P260-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
Precautionary Statement H290-H314-H335
EINECSNumber 231-595-7
RTECSNumber MW4025000