missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, ACS Reagent, 37%, Honeywell Fluka™
ACS Reagent, 37%
Supplier: Honeywell Inc 2581484L
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
| Hydrochloric Acid | |
| 37% | |
| ≤5ppm | |
| 4 L | |
| MFCD00011324 MFCD00792839 | |
| 1098214 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| hydrogen chloride | |
| 313 | |
| 36.46g/mol |
| 7647-01-0 | |
| 1.2 g/mL at 25°C | |
| 3.23psi (21.1°C), 7.93psi (37.7°C) | |
| ClH | |
| 1789 | |
| hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
| Cl | |
| 36.46 | |
| CHEBI:17883 | |
| ACS Reagent |
Chemical Identifiers
| 7647-01-0 | |
| 36.46 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| 313 | |
| Cl |
| ClH | |
| MFCD00011324 MFCD00792839 | |
| hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
| CHEBI:17883 |
Safety and Handling
P260-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
H290-H314-H335
EINECSNumber : 231-595-7
RTECSNumber : MW4025000