missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 6.25 N, Ricca Chemical

Catalog No. p-3251789
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Amber Glass Bottle
Natural Poly Bottle
Safety-coated Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
375532G 1 L Amber Glass Bottle
375532 1 L Natural Poly Bottle
37555PT 2.5 L Safety-coated Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 375532G Supplier Ricca Chemical Company Supplier No. 375532G
Only null left

Chemical Identifiers

CAS 7732-18-5 , 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

Color Colorless to Green-Yellow
Concentration 6.25Normal
CAS Min % 61.0, 37.21
CAS Max % 63.0, 38.73
Packaging Amber Glass Bottle
Quantity 1 L
Physical Form Liquid
Grade Laboratory
Chemical Name or Material Hydrochloric Acidl