missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, 6.25 N, Ricca Chemical
Supplier: Ricca Chemical Company 37555PT
Specifications
| Hydrochloric Acidl | |
| 7732-18-5,7647-01-0 | |
| 6.25Normal | |
| Safety-coated Glass Bottle | |
| ClH | |
| MFCD00011324 MFCD00792839 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| hydrogen chloride | |
| 313 | |
| Laboratory |
| Colorless to Green-Yellow | |
| 61.0,37.21 | |
| 63.0,38.73 | |
| 2.5 L | |
| Liquid | |
| hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
| Cl | |
| 36.46 | |
| CHEBI:17883 |
Chemical Identifiers
| 7732-18-5 | |
| 36.46 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| 313 | |
| hydrogen chloride |
| ClH | |
| MFCD00011324 MFCD00792839 | |
| hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
| CHEBI:17883 | |
| Cl |