missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 6.0 N, Aqueous Solution, APHA, Spectrum™ Chemical

Catalog No. 18610924
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 L
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610924 1 L Poly Bottle
18610925 2.5 L Poly Bottle
18610926 4 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18610924 Supplier Spectrum Chemical Mfg Cor Supplier No. H1601LT
Only null left

Chemical Identifiers

CAS 7647-01-0 , 7732-18-5

Specifications

Concentration 6.0 N ± 0.2 N
Packaging Poly Bottle
Quantity 1 L
Grade APHA/EPA
Chemical Name or Material Hydrochloric Acid

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.