missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 50% (v/v), Ricca Chemical

Catalog No. R3580000100
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
10 L
Packaging:
Cubitainer
Natural Poly Bottle
Safety-coated Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R3580000100 100 mL Natural Poly Bottle
358016 500 mL Natural Poly Bottle
358032 1 L Natural Poly Bottle
35805PT 2.5 L Safety-coated Glass Bottle
358025 10 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. R3580000100 Supplier Ricca Chemical Company Supplier No. R3580000100A
Only null left

Chemical Identifiers

CAS 7732-18-5 , 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

Color Colorless
Odor Slightly pungent to pungent
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Assay Percent Range 50.0 ± 0.4% (v/v)
Quantity 100 mL
Physical Form Liquid
Solubility Information Solubility in Water: Infinite
Percent Purity 50% V/V
Grade Laboratory
Chemical Name or Material Hydrochloric Acid, 50% (v/v)
Show More Show Less