missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid 37% w/w, ACS Reagent, Reagents

Catalog No. p-200020932
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
20 L
Packaging:
Glass Bottle
HDPE Winpak
Poly Bottle
Safety Coated Glass
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01336840 1 L Glass Bottle
01336839 2.5 L Poly Bottle
01336838 2.5 L Safety Coated Glass
01336837 2.5 L Glass Bottle
01336836 20 L HDPE Winpak
01336835 4 L Poly Bottle
01336834 500 mL Poly Bottle
01336833 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 01336840 Supplier Labchem Inc Supplier No. C51011001C
Only null left

Hydrochloric Acid 37% w/w, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7647-01-0
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Muriatic Acid, Chlorohydric Acid
IUPAC Name water
SMILES O

Specifications

pH <1
Color Colorless to Slightly Greenish-Yellow
Boiling Point 100°C
Packaging Glass Bottle
Specific Gravity 1.18
Quantity 1 L
Physical Form Liquid
Grade ACS
Chemical Name or Material Hydrochloric Acid 37% w/w, ACS Reagent