missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid 37% w/w, ACS Reagent, Reagents
Hydrochloric Acid 37% w/w, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C510110025D
Specifications
| <1 | |
| Colorless to Slightly Greenish-Yellow | |
| 100°C | |
| 1.18 | |
| H2O | |
| Muriatic Acid, Chlorohydric Acid | |
| O | |
| 18.02 |
| Hydrochloric Acid 37% w/w, ACS Reagent | |
| 7732-18-5,7647-01-0 | |
| Safety Coated Glass | |
| 2.5 L | |
| Liquid | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water | |
| ACS |
Chemical Identifiers
| 7732-18-5 | |
| 18.02 | |
| Muriatic Acid, Chlorohydric Acid | |
| O |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water |