missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, 30.0%, Suprapur™, MilliporeSigma™
$283.07 - $283.07
Chemical Identifiers
| CAS | 7647-01-0 |
|---|---|
| Molecular Formula | ClH |
| Molecular Weight (g/mol) | 36.46 |
| MDL Number | MFCD00011324 MFCD00792839 |
| InChI Key | VEXZGXHMUGYJMC-UHFFFAOYSA-N |
| Synonym | Hydrogen chloride solution |
| IUPAC Name | hydrogen chloride |
| SMILES | Cl |
Chemical Identifiers
| 7647-01-0 | |
| 36.46 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| hydrogen chloride |
| ClH | |
| MFCD00011324 MFCD00792839 | |
| Hydrogen chloride solution | |
| Cl |
Specifications
| < 1 (300Â g/L, H2O, 20°C) | |
| 7647-01-0 | |
| 1.15Â g/cm3 (at 20°C) | |
| 21.8Â hPa (20°C) | |
| 250 mL | |
| MFCD00011324 MFCD00792839 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| hydrogen chloride |
| −50°C | |
| 85°C (1013Â hPa) | |
| Plastic Bottle | |
| ≥ 30.0% | |
| ClH | |
| Hydrogen chloride solution | |
| Cl | |
| 36.46 |