missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 30.0%, Suprapur™, MilliporeSigma™

Catalog No. 361044622
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
361044622 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 361044622 Supplier Technidata Supplier No. 1.00318.0250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym Hydrogen chloride solution
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

pH < 1 (300 g/L, H2O, 20°C)
Melting Point −50°C
Boiling Point 85°C (1013 hPa)
Density 1.15 g/cm3 (at 20°C)
Packaging Plastic Bottle
Vapor Pressure 21.8 hPa (20°C)
Assay Percent Range ≥ 30.0%
Quantity 250 mL
Percent Purity 30%
Grade Not Available
Chemical Name or Material Hydrochloric Acid
Show More Show Less