missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 3 N Solution, Spectrum™ Chemical

Catalog No. p-7165667
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610903 500 mL Amber Glass Bottle
18610904 1 L Amber Glass Bottle
18610906 4 L Amber Glass Bottle
18610905 20 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18610903 Supplier Spectrum Chemical Mfg Cor Supplier No. H150500ML
Only null left

Chemical Identifiers

CAS 7647-01-0 , 7732-18-5

Specifications

Concentration 3.00 N ± 0.03 N
CAS Max % 10.4%, 89.6%
Packaging Amber Glass Bottle
Quantity 500 mL
Grade Ungraded
Chemical Name or Material Hydrochloric Acid

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.