missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, 3.00 N, Ricca Chemical
$76.40 - $800.37
Chemical Identifiers
| CAS | 7732-18-5 |
|---|---|
| Molecular Formula | ClH |
| Molecular Weight (g/mol) | 36.46 |
| MDL Number | MFCD00011324 MFCD00792839 |
| InChI Key | VEXZGXHMUGYJMC-UHFFFAOYSA-N |
| Synonym | hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof |
| PubChem CID | 313 |
| ChEBI | CHEBI:17883 |
| IUPAC Name | hydrogen chloride |
| SMILES | Cl |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
3720-16
|
Ricca Chemical Company
372016 |
500 mL | Natural Poly Bottle |
Each for $76.40
|
|
||||
|
3720-32
|
Ricca Chemical Company
372032 |
1 L | Natural Poly Bottle |
Each for $124.40
|
|
||||
|
3720-1
|
Ricca Chemical Company
37201 |
4 L | Natural Poly Bottle |
Each for $372.13
|
|
||||
|
3720-25
|
Ricca Chemical Company
372025 |
10 L | Cubitainer |
Each for $598.63
|
|
||||
|
3720-5
|
Ricca Chemical Company
37205 |
20 L | Cubitainer |
Each for $800.37
|
|
||||
Chemical Identifiers
| 7732-18-5 | |
| 36.46 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| 313 | |
| hydrogen chloride |
| ClH | |
| MFCD00011324 MFCD00792839 | |
| hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
| CHEBI:17883 | |
| Cl |
Specifications
| Colorless to Green-Yellow | |
| 76.0,22.32 | |
| 79.0,23.24 | |
| 500 mL | |
| Liquid | |
| hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
| Cl | |
| 36.46 | |
| CHEBI:17883 | |
| Hydrochloric Acid |
| 7732-18-5,7647-01-0 | |
| 3.00Normal | |
| Natural Poly Bottle | |
| ClH | |
| MFCD00011324 MFCD00792839 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| hydrogen chloride | |
| 313 | |
| Laboratory |