missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 3.00 N, Ricca Chemical

Catalog No. 372016
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
372016 500 mL Natural Poly Bottle
372032 1 L Natural Poly Bottle
37201 4 L Natural Poly Bottle
372025 10 L Cubitainer
37205 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 372016 Supplier Ricca Chemical Company Supplier No. 372016
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7732-18-5 | Formula: ClH | Weight: 36.46 g/mol

Chemical Identifiers

CAS 7732-18-5 , 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

Color Colorless to Green-Yellow
CAS Min % 76.0, 22.32
CAS Max % 79.0, 23.24
Concentration 3.00Normal
Packaging Natural Poly Bottle
Quantity 500 mL
Physical Form Liquid
Grade Not Available
Chemical Name or Material Hydrochloric Acid