missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, 3.00 N, Electronic Grade, Ricca Chemical
$591.50 - $591.50
Chemical Identifiers
| CAS | 7732-18-5 |
|---|---|
| Molecular Formula | ClH |
| Molecular Weight (g/mol) | 36.46 |
| MDL Number | MFCD00011324 MFCD00792839 |
| InChI Key | VEXZGXHMUGYJMC-UHFFFAOYSA-N |
| Synonym | hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof |
| PubChem CID | 313 |
| ChEBI | CHEBI:17883 |
| IUPAC Name | hydrogen chloride |
| SMILES | Cl |
Chemical Identifiers
| 7732-18-5 | |
| 36.46 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| 313 | |
| hydrogen chloride |
| ClH | |
| MFCD00011324 MFCD00792839 | |
| hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
| CHEBI:17883 | |
| Cl |
Specifications
| Colorless to Green-Yellow | |
| 76.0,22.19 | |
| Natural Poly Bottle | |
| ClH | |
| MFCD00011324 MFCD00792839 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| hydrogen chloride | |
| 313 | |
| Electronic | |
| Hydrochloric Acid |
Safety and Handling
ShelfLife : 18 months