missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 20° Be, Technical, Approx. 30%, Spectrum™ Chemical

H1040, 7647-01-0, HCl

$423.53 - $5752.83

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
IUPAC Name hydrogen chloride
SMILES Cl
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18603175
View Documents
Spectrum Chemical Mfg Cor
H10404LTPV
4 L Poly-Coated Glass Bottle
Each for $423.53
Only null left
 
18603176
View Documents
Spectrum Chemical Mfg Cor
H10404LTPVCS4
4 x 4 L Poly-Coated Glass Bottle
Case of 4 Each for $1,065.17
Only null left
 
18603177
View Documents
Spectrum Chemical Mfg Cor
H104020LT
20 L Poly Drum
Each for $1,275.90
Only null left
 
18603178
View Documents
Spectrum Chemical Mfg Cor
H104055GLBL
55 gal. Drum
Each for $5,752.83
Only null left
 
Description

Description

Spectrum™ Chemical Hydrochloric Acid, 20 DEG Be, Technical is a colorless and clear, highly corrosive, highly pungent solution of hydrogen chloride in water. As an monoprotic acid and one of the six common strong mineral acids it is the least likely to be subject to an oxidation-reduction reaction. Hydrochloric acid is also frequently used during chemical analysis to prepare samples for analysis. It reacts with basic compounds, turning them into dissolved chlorides that can then undergo analysis.
Specifications

Chemical Identifiers

7647-01-0
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
Cl
ClH
MFCD00011324 MFCD00792839
hydrogen chloride

Specifications

7647-01-0
Poly-Coated Glass Bottle
ClH
UN1789
Cl
36.46
Technical
100%
4 L
MFCD00011324 MFCD00792839
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrogen chloride
∼30%
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'