missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 20° Be, Technical, Approx. 30%, Spectrum™ Chemical

H1040, 7647-01-0, HCl

Supplier:  Spectrum Chemical Mfg Cor H104020LT

 View more versions of this product

Catalog No. 18603177

Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Hydrochloric Acid, 20 DEG Be, Technical is a colorless and clear, highly corrosive, highly pungent solution of hydrogen chloride in water. As an monoprotic acid and one of the six common strong mineral acids it is the least likely to be subject to an oxidation-reduction reaction. Hydrochloric acid is also frequently used during chemical analysis to prepare samples for analysis. It reacts with basic compounds, turning them into dissolved chlorides that can then undergo analysis.
Specifications

Specifications

7647-01-0
Poly Drum
ClH
UN1789
Cl
36.46
Technical
100%
20 L
MFCD00011324 MFCD00792839
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrogen chloride
∼30%

Chemical Identifiers

7647-01-0
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
Cl
ClH
MFCD00011324 MFCD00792839
hydrogen chloride
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions