missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 2.50 N, Ricca Chemical

Supplier:  Ricca Chemical Company R3715000500A

 View more versions of this product

Catalog No. R3715000500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

Hydrochloric Acid
79.0,19.31
Natural Poly Bottle
ClH
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
[H+].[Cl-]
36.46
CHEBI:17883
7647-01-0 , 7732-18-5
82.0,20.1
500 mL
MFCD00011324
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrogen chloride
313

Chemical Identifiers

7647-01-0
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
hydrogen chloride
ClH
MFCD00011324
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
CHEBI:17883
[H+].[Cl-]
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions