missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, 2.50 Normal, Reagents
Specifications
Specifications
| pH | <1 |
| Color | Colorless to Green/Yellow |
| Boiling Point | 100°C |
| Packaging | Poly Bottle |
| Specific Gravity | 1.0433 |
| Quantity | 100 mL |
| Physical Form | Liquid |
| Grade | Reagent |
| Chemical Name or Material | Hydrochloric Acid, 2.50 Normal |