missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 2.50 Normal, Reagents

Catalog No. 01335014
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335014 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01335014 Supplier Labchem Inc Supplier No. CH123500100A
Only null left

Hydrochloric Acid, 2.50 Normal, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 7732-18-5 , 7647-01-0
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Hydrogen chloride solution
IUPAC Name water
SMILES O

Specifications

pH <1
Color Colorless to Green/Yellow
Boiling Point 100°C
Packaging Poly Bottle
Specific Gravity 1.0433
Quantity 100 mL
Physical Form Liquid
Grade Reagent
Chemical Name or Material Hydrochloric Acid, 2.50 Normal