missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, 2.00 N, Ricca Chemical
Specifications
Specifications
| Color | Colorless |
| Concentration | 2.00N |
| CAS Min % | 16.05, 82.0 |
| CAS Max % | 16.71, 85.0 |
| Packaging | Natural Poly Bottle |
| Quantity | 500 mL |
| Physical Form | Liquid |
| Grade | Laboratory |
| Chemical Name or Material | Hydrochloric Acid |