missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 2.0 N Solution, 2 N +/-0.08 N, Spectrum™ Chemical

Catalog No. 18603174
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603174 20 L Poly Bottle
18603172 1 L Poly Bottle
18603173 4 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18603174 Supplier Spectrum Chemical Mfg Cor Supplier No. H14520LT
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

H-145, 7647-01-0

Spectrum™ Chemical Hydrochloric Acid, 2.0 N Solution is a colorless, clear, highly corrosive, highly pungent solution of hydrogen chloride in water. As an monoprotic acid and one of the six common strong mineral acids it is the least likely to be subject to an oxidation-reduction reaction. Hydrochloric acid is frequently used during chemical analysis to prepare samples for analysis. It reacts with basic compounds, turning them into dissolved chlorides that can then undergo analysis.

Chemical Identifiers

CAS 7647-01-0 , 7732-18-5
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

Concentration 2 N ±0.08
CAS Max % 7 to 7.5%, 92.5 to 93%
Packaging Poly Bottle
Quantity 20 L
UN Number UN1789
Grade Ungraded
Chemical Name or Material Hydrochloric Acid