missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric acid, 1N volumetric Standard solution, Macron Fine Chemicals™
macron
Chemical Identifiers
| CAS | 7647-01-0 |
|---|---|
| Molecular Formula | ClH |
| Molecular Weight (g/mol) | 36.46 |
| MDL Number | MFCD00011324 |
| InChI Key | VEXZGXHMUGYJMC-UHFFFAOYSA-N |
| Synonym | hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof |
| PubChem CID | 313 |
| ChEBI | CHEBI:17883 |
| IUPAC Name | hydrogen chloride |
| SMILES | [H+].[Cl-] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
MK638820
|
Avantor Macron
638805 |
20L | Cubitainer |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| ClH | |
| MFCD00011324 | |
| hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
| CHEBI:17883 | |
| [H+].[Cl-] |
Specifications
| 7647-01-0 | |
| MFCD00011324 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| hydrogen chloride | |
| 313 | |
| 36.46 |
| ClH | |
| hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
| [H+].[Cl-] | |
| 36.46 | |
| CHEBI:17883 | |
| Hydrochloric acid, 1N volumetric standard solution |