missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric acid, 1N volumetric Standard solution, Macron Fine Chemicals™
macron
Supplier: Avantor 638805
| Quantity | 20L |
|---|---|
| Packaging | Cubitainer |
Chemical Identifiers
| ClH | |
| MFCD00011324 | |
| hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
| CHEBI:17883 | |
| [H+].[Cl-] |
Specifications
| Hydrochloric acid, 1N volumetric standard solution | |
| Cubitainer | |
| ClH | |
| hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
| [H+].[Cl-] | |
| 36.46 | |
| CHEBI:17883 |
| 7647-01-0 | |
| 20L | |
| MFCD00011324 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| hydrogen chloride | |
| 313 | |
| 36.46 |