missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 12.0 N, Ricca Chemical

Catalog No. p-7239399
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Natural Poly Bottle
Safety-Coated Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R37800001A 1 L Natural Poly Bottle
37805PT 2.5 L Safety-Coated Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R37800001A Supplier Ricca Chemical Company Supplier No. R37800001A
Only null left

Chemical Identifiers

CAS 7732-18-5 , 7647-01-0

Specifications

Appearance of Solution Colorless to slightly Greenish-yellow Liquid
Concentration 11.8 to 12.2 N
CAS Max % 63, 37
Packaging Natural Poly Bottle
Quantity 1 L
Molecular Formula H2O
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILES O
IUPAC Name water
Molecular Weight (g/mol) 36.46 g/mol
Grade Laboratory
Assay Normal at 20°C
Chemical Name or Material Hydrochloric Acid
Show More Show Less
ShelfLife 24 months