Learn More
Hydrochloric acid, 10% v/v aq. soln.
CAS: 7647-01-0 | ClH | 36.46 g/mol
$55.08 - $129.75
Chemical Identifiers
| CAS | 7647-01-0 |
|---|---|
| Molecular Formula | ClH |
| Molecular Weight (g/mol) | 36.46 |
| MDL Number | MFCD00011324 MFCD00792839 |
| InChI Key | VEXZGXHMUGYJMC-UHFFFAOYSA-N |
| Synonym | hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof |
| PubChem CID | 313 |
| ChEBI | CHEBI:17883 |
| SMILES | Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA35611AP
|
Thermo Scientific Chemicals
035611AP |
500 mL |
Each for $55.08
|
|
|||||
|
AA35611K2
|
Thermo Scientific Chemicals
035611K2 |
1 L |
Each for $76.44
|
|
|||||
|
AA35611K7
|
Thermo Scientific Chemicals
035611K7 |
4 L |
Each for $129.75
|
|
|||||
Description
Hydrochloric acid is used in chemical synthesis reactions to produce vinyl chloride, fertilizers, dyes, pigments and pharmaceutical ingredients. It is also used in catalyst regeneration, rubber manufacturing, electroplating, semiconductor manufacturing, leather tanning and cotton seed processing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7647-01-0 | |
| 36.46 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| 313 | |
| Cl |
| ClH | |
| MFCD00011324 MFCD00792839 | |
| hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
| CHEBI:17883 |
Specifications
| Colorless to pale yellow liquid | |
| 7647-01-0,7732-18-5 | |
| 500 mL | |
| Liquid | |
| hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| chlorane | |
| 313 | |
| 36.46 | |
| Hydrochloric acid |
| Colorless to Yellow | |
| 23 hPa (17mm Hg) at 20°C | |
| ClH | |
| MFCD00011324 MFCD00792839 | |
| Completely miscible with water. | |
| Cl | |
| 36.46 | |
| CHEBI:17883 | |
| Hydrochloric acid: 10.0%; Water: 90.0% |
Safety and Handling
P234-P264b-P280-P302+P352-P305+P351+P338-P310-P332+P313-P362-P390
H290-H315-H318
DOTInformation : Hazard Class: 8, Packing Group: III
EINECSNumber : 231-595-7
RTECSNumber : MW4025000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only