missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric acid, 10% v/v aq. soln.

CAS: 7647-01-0 | ClH | 36.46 g/mol

Supplier:  Thermo Scientific Chemicals 035611K7

 View more versions of this product

Catalog No. AA35611K7


Only null left
Explore available promotions

Description

Description

Hydrochloric acid is used in chemical synthesis reactions to produce vinyl chloride, fertilizers, dyes, pigments and pharmaceutical ingredients. It is also used in catalyst regeneration, rubber manufacturing, electroplating, semiconductor manufacturing, leather tanning and cotton seed processing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Colorless to pale yellow liquid
Colorless to Yellow
23 hPa (17mm Hg) at 20°C
ClH
MFCD00011324 MFCD00792839
Completely miscible with water.
Cl
36.46
CHEBI:17883
Hydrochloric acid: 10.0%; Water: 90.0%
Hydrochloric acid
7647-01-0,7732-18-5
4 L
Liquid
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
VEXZGXHMUGYJMC-UHFFFAOYSA-N
chlorane
313
36.46

Chemical Identifiers

7647-01-0
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
Cl
ClH
MFCD00011324 MFCD00792839
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
CHEBI:17883
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P234-P264b-P280-P302+P352-P305+P351+P338-P310-P332+P313-P362-P390

H290-H315-H318

DOTInformation : Hazard Class: 8, Packing Group: III

EINECSNumber : 231-595-7

RTECSNumber : MW4025000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only