missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 10.0 N Solution, Spectrum™ Chemical

Catalog No. p-7165686
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610890 1 L Poly Bottle
18610891 4 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18610890 Supplier Spectrum Chemical Mfg Cor Supplier No. H2851LT
Only null left

Chemical Identifiers

CAS 7647-01-0 , 7732-18-5

Specifications

Concentration 10 N ±0.5 N
Packaging Poly Bottle
Quantity 1 L
Grade Ungraded
Chemical Name or Material Hydrochloric Acid

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.