missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 1.9328 Normal, Reagents

Catalog No. p-200021343
Change view
Click to view available options
Quantity:
1 L
10 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334991 10 L
01334990 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01334991 Supplier Labchem Inc Supplier No. CH15150010E2
Only null left

Hydrochloric Acid, 1.9328 Normal, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 7732-18-5 , 7647-01-0
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Hydrogen chloride solution
IUPAC Name water
SMILES O

Specifications

pH <1
Color Colorless
Boiling Point 100°C
Packaging HDPE Winpak
Specific Gravity 1.0337
Quantity 10 L
Physical Form Liquid
Grade Reagent
Chemical Name or Material Hydrochloric Acid, 1.9328 Normal