missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 1.50 Normal, Reagents

Catalog No. 01335017
Change view
Click to view available options
Quantity:
20 L
200 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335017 200 L
01335016 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01335017 Supplier Labchem Inc Supplier No. CH122500200E

Hydrochloric Acid, 1.50 Normal, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 7732-18-5 , 7647-01-0
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Hydrogen chloride solution
IUPAC Name water
SMILES O

Specifications

pH <1
Color Colorless
Boiling Point 100°C
Packaging Poly Drum
Specific Gravity 1.0263
Quantity 200 L
Physical Form Liquid
Grade Reagent
Chemical Name or Material Hydrochloric Acid, 1.50 Normal