missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid (1+3), Fisher Chemical™
Specifications
Specifications
| CAS Min % | 76.0, 22.32 |
| CAS Max % | 79.0, 23.24 |
| Packaging | Natural Poly Bottle |
| Quantity | 1 L |
| Percent Purity | 25% V/V |
| Grade | Laboratory |
| Chemical Name or Material | Hydrochloric Acid (1+3) |
Safety and Handling
| DOTInformation | DOT Class 8, : Corrosive |