missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, 1:1, 50% v/v Approximately 6N, Reagents
Hydrochloric Acid, 1:1, 50% v/v Approximately 6N, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$119.40 - $1151.40
Chemical Identifiers
| CAS | 7732-18-5 |
|---|---|
| Molecular Formula | H2O |
| Molecular Weight (g/mol) | 18.02 |
| InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
| Synonym | Hydrogen chloride solution |
| IUPAC Name | water |
| SMILES | O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
01-336-789
|
Labchem Inc
C550500010F |
10 L |
Each for $576.23
|
|
|||||
|
01-336-788
|
Labchem Inc
C55050001A |
1 L |
Each for $161.70
|
|
|||||
|
01-336-787
|
Labchem Inc
C550500025D1 |
2.5 L |
Each for $226.80
|
|
|||||
|
01-336-786
|
Labchem Inc
C550500020E7 |
20 L |
Each for $1,151.40
|
|
|||||
|
01-336-785
|
Labchem Inc
C55050004A |
4 L |
Each for $405.90
|
|
|||||
|
01-336-784
|
Labchem Inc
C5505000500A |
500 mL |
Each for $119.40
|
|
|||||
Chemical Identifiers
| 7732-18-5 | |
| 18.02 | |
| Hydrogen chloride solution | |
| O |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water |
Specifications
| 0 | |
| 7732-18-5,7647-01-0 | |
| Cubitainer | |
| 10 L | |
| Liquid | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water | |
| 50% | |
| Hydrochloric Acid, 1:1, 50% v/v Approximately 6N |
| Colorless to Slight Yellow | |
| 108°C | |
| 1.0999 | |
| H2O | |
| Hydrogen chloride solution | |
| O | |
| 18.02 | |
| Reagent |