missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, 1:1, 50% v/v Approximately 6N, Reagents
Hydrochloric Acid, 1:1, 50% v/v Approximately 6N, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C550500025D1
Specifications
| 0 | |
| Colorless to Slight Yellow | |
| 108°C | |
| 1.0999 | |
| H2O | |
| Hydrogen chloride solution | |
| O | |
| 18.02 | |
| Reagent |
| Hydrochloric Acid, 1:1, 50% v/v Approximately 6N | |
| 7732-18-5,7647-01-0 | |
| Glass Bottle | |
| 2.5 L | |
| Liquid | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water | |
| 50% |
Chemical Identifiers
| 7732-18-5 | |
| 18.02 | |
| Hydrogen chloride solution | |
| O |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water |