Learn More
Hydrochloric acid, 1.0N Standardized Solution
CAS: 7647-01-0 | ClH | 36.46 g/mol
Supplier: Thermo Scientific Chemicals 035640K7
Description
Hydrochloric acid, 1.0N standardized solution is used for volumetric analysis in laboratories.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Hydrochloric acid | |
| 23 hPa (17mm Hg) at 20°C | |
| ClH | |
| Colorless to Yellow | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| chlorane | |
| 4 L | |
| CHEBI:17883 | |
| Hydrochloric acid: 8.26%; Water: 91.74% |
| 7647-01-0,7732-18-5 | |
| Liquid | |
| MFCD00011324 MFCD00792839 | |
| Miscible with water, methanol,ethanol and ether | |
| Cl | |
| 36.46 | |
| 313 | |
| 36.46 |
Chemical Identifiers
| 7647-01-0 | |
| 36.46 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| CHEBI:17883 |
| ClH | |
| MFCD00011324 MFCD00792839 | |
| 313 | |
| Cl |
Safety and Handling
P234-P264b-P280-P302+P352-P305+P351+P338-P310-P332+P313-P362-P390
H290-H315-H318
DOTInformation : Hazard Class: 8; Packaging Group: III
EINECSNumber : 231-595-7
RTECSNumber : MW4025000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only