missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 1.00 N, Ricca Tightrant™

Catalog No. 370032
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
370032 1 L Natural Poly Bottle
370016 500 mL Natural Poly Bottle
37001CT 4 L Cubitainer
37001 4 L Natural Poly Bottle
370025 10 L Cubitainer
37005 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 370032 Supplier Ricca Chemical Company Supplier No. RI370032
Edge
  • Tested in and ISO 17025 Accredited Facility
  • Test Reports provided with Certified Values and Measurement Uncertainty
  • Tight release specifications for low lot-to-lot variability

Chemical Identifiers

CAS 7732-18-5 , 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

CAS Min % 89.0, 8.71
CAS Max % 93.0, 9.07
Packaging Natural Poly Bottle
Quantity 1 L
Grade Laboratory
Concentration or Composition (by Analyte or Components) 1.00N
Chemical Name or Material Hydrochloric Acid