missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, 1.00 Normal, Reagents
Hydrochloric Acid, 1.00 Normal, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. Accredited to ISO 17025, NIST Traceable.
$72.13 - $2142.17
Chemical Identifiers
| CAS | 7732-18-5 |
|---|---|
| Molecular Formula | H2O |
| Molecular Weight (g/mol) | 18.02 |
| InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
| Synonym | Hydrogen chloride solution |
| IUPAC Name | water |
| SMILES | O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
01-336-598
|
Labchem Inc
C710108010F |
10 L | Cubitainer |
Each for $385.37
|
|
||||
|
01-336-597
|
Labchem Inc
C71010801A |
1 L | Poly Bottle |
Each for $81.63
|
|
||||
|
01-336-596
|
Labchem Inc
C7101080200E |
200 L | Poly Drum |
Each for $2,142.17
|
|
||||
|
01-336-595
|
Labchem Inc
C710108020E3 |
20 L |
Each for $496.97
|
|
|||||
|
01-336-594
|
Labchem Inc
C710108020F |
20 L | Cubitainer |
Each for $496.97
|
|
||||
|
01-336-593
|
Labchem Inc
C71010804A |
4 L | Poly Bottle |
Each for $142.47
|
|
||||
|
01-336-592
|
Labchem Inc
C71010804F |
4 L | Cubitainer |
Each for $180.80
|
|
||||
|
01-336-591
|
Labchem Inc
C7101080500A |
500 mL | Poly Bottle |
Each for $72.13
|
|
||||
Chemical Identifiers
| 7732-18-5 | |
| 18.02 | |
| Hydrogen chloride solution | |
| O |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water |
Specifications
| <1 | |
| 7732-18-5,7647-01-0 | |
| Cubitainer | |
| 10 L | |
| Liquid | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water | |
| Reagent |
| Colorless | |
| 100°C | |
| 1.0177 | |
| H2O | |
| Hydrogen chloride solution | |
| O | |
| 18.02 | |
| Hydrochloric Acid, 1.00 Normal |