missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, 1.00 Normal, Reagents
Hydrochloric Acid, 1.00 Normal, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. Accredited to ISO 17025, NIST Traceable.
Supplier: Labchem Inc C71010804F
Specifications
| <1 | |
| Colorless | |
| 100°C | |
| 1.0177 | |
| H2O | |
| Hydrogen chloride solution | |
| O | |
| 18.02 |
| Hydrochloric Acid, 1.00 Normal | |
| 7732-18-5,7647-01-0 | |
| Cubitainer | |
| 4 L | |
| Liquid | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water | |
| Reagent |
Chemical Identifiers
| 7732-18-5 | |
| 18.02 | |
| Hydrogen chloride solution | |
| O |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water |