missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 0.600 Normal, Reagents

Catalog No. 01335102
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
200 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335102 20 L
01335104 1 L
01335103 200 L
01335101 4 L
01335100 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01335102 Supplier Labchem Inc Supplier No. CH10300020F

Hydrochloric Acid, 0.600 Normal, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7647-01-0
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Hydrogen chloride solution
IUPAC Name water
SMILES O

Specifications

pH <1
Color Colorless
Boiling Point 100°C
Packaging Cubitainer
Specific Gravity 1.0107
Quantity 20 L
Physical Form Liquid
Grade Reagent
Chemical Name or Material Hydrochloric Acid, 0.600 Normal