missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, 0.600 N, Ricca Chemical
Specifications
Specifications
| CAS Min % | 93.0, 5.44 |
| CAS Max % | 96.0, 5.66 |
| Packaging | Natural Poly Bottle |
| Quantity | 1 L |
| Grade | Laboratory |
| Chemical Name or Material | Hydrochloric Acid |