missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 0.600 N, Ricca Chemical

Catalog No. 365532
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
365532 1 L Natural Poly Bottle
36551 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 365532 Supplier Ricca Chemical Company Supplier No. 365532
Only null left

Chemical Identifiers

CAS 7732-18-5 , 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

CAS Min % 93.0, 5.44
CAS Max % 96.0, 5.66
Packaging Natural Poly Bottle
Quantity 1 L
Grade Laboratory
Chemical Name or Material Hydrochloric Acid