missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 0.5167 Normal, Reagents

Catalog No. 01335114
Change view
Click to view available options
Quantity:
1 L
4 L
10 L
20 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335114 10 L
01335113 1 L
01335112 20 L
01335111 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01335114 Supplier Labchem Inc Supplier No. CH10230010F

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Hydrochloric Acid, 0.5167 Normal, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 7732-18-5 , 7647-01-0
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Hydrogen chloride solution
IUPAC Name water
SMILES O

Specifications

pH <1
Color Colorless
Boiling Point 100°C
Packaging Cubitainer
Specific Gravity 1.0092
Quantity 10 L
Physical Form Liquid
Grade Reagent
Chemical Name or Material Hydrochloric Acid, 0.5167 Normal